ENA408616017 Enamine

6-chloro-3-(chloromethyl)-2-(trifluoromethyl)pyridine

  • Molecular Weight : 230.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-3-(chloromethyl)-2-(trifluoromethyl)pyridine
Inchi Code 1S/C7H4Cl2F3N/c8-3-4-1-2-5(9)13-6(4)7(10,11)12/h1-2H,3H2
Inchi Key FQIAUXNJSLDTPL-UHFFFAOYSA-N

Description

Physical Form liquid
Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136990
Availability details for
Quantity
  • Availability
  • Add to Cart