ENA408616018 Enamine

3-chloro-N-cyclopropyl-6-fluoro-1-benzothiophene-2-carboxamide

  • MDL number MFCD02711679
  • Cas Number : 438025-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137020
Availability details for
Quantity
  • Availability
  • Add to Cart