ENA408616019 Enamine

5-fluoro-N-(1H-pyrazol-4-yl)-1-benzothiophene-2-carboxamide

  • MDL number MFCD12075468
  • Cas Number : 1153800-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-137021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137021
Availability details for
Quantity
  • Availability
  • Add to Cart