ENA408616020 Enamine

2-[(2,3-dimethylphenoxy)methyl]benzoic acid

  • MDL number MFCD09707344
  • Cas Number : 938143-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137022
Availability details for
Quantity
  • Availability
  • Add to Cart