ENA408616022 Enamine

2-[(2,3-dichlorophenoxy)methyl]benzoic acid

  • MDL number MFCD09704513
  • Cas Number : 938124-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137024
Availability details for
Quantity
  • Availability
  • Add to Cart