ENA408616023 Enamine

N-ethyl-4-(4-fluorophenyl)piperazine-1-carboxamide

  • MDL number MFCD08885619
  • Cas Number : 923121-38-4
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137025
Availability details for
Quantity
  • Availability
  • Add to Cart