ENA408616024 Enamine

2-{[(4-hydroxy-6-propylpyrimidin-2-yl)sulfanyl]methyl}furan-3-carboxylic acid

  • MDL number MFCD26407790
  • Cas Number : 1153555-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137027

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137027
Availability details for
Quantity
  • Availability
  • Add to Cart