ENA408616026 Enamine

4-fluoro-3-(methoxymethyl)-1-benzothiophene-2-carboxamide

  • MDL number MFCD11769086
  • Cas Number : 1089555-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137029
Availability details for
Quantity
  • Availability
  • Add to Cart