ENA408616027 Enamine

3-{[(2-methylpropoxy)carbonyl]amino}naphthalene-2-carboxylic acid

  • MDL number MFCD13355815
  • Cas Number : 221070-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137031

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137031
Availability details for
Quantity
  • Availability
  • Add to Cart