ENA408616036 Enamine

2-(5-methylpyrazin-2-yl)ethan-1-ol

  • MDL number MFCD22413827
  • Cas Number : 142780-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133145

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133145
Availability details for
Quantity
  • Availability
  • Add to Cart