ENA408616037 Enamine

N-cyclohexylhydrazinecarbothioamide

  • MDL number MFCD00021318
  • Cas Number : 21198-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03774
Availability details for
Quantity
  • Availability
  • Add to Cart