ENA408616039 Enamine

(4-fluorophenyl)[4-(trifluoromethyl)phenyl]methanamine

  • MDL number MFCD09931988
  • Cas Number : 1016671-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133203

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133203
Availability details for
Quantity
  • Availability
  • Add to Cart