ENA408616046 Enamine

2-[4-(aminomethyl)phenyl]-1$l^{6},2-thiazolidine-1,1-dione

  • MDL number MFCD09805348
  • Cas Number : 1016529-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137132

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137132
Availability details for
Quantity
  • Availability
  • Add to Cart