ENA408616050 Enamine

N-hydroxydecanamide

  • MDL number MFCD01736048
  • Cas Number : 2259-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137136
Availability details for
Quantity
  • Availability
  • Add to Cart