ENA408616053 Enamine

3-{[4-(difluoromethyl)phenyl]methyl}pyrrolidine

  • MDL number MFCD24225542
  • Cas Number : 1337424-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137139
Availability details for
Quantity
  • Availability
  • Add to Cart