ENA408616064 Enamine

methyl 2-(5-bromo-1,3-thiazol-2-yl)acetate

  • MDL number MFCD14706346
  • Cas Number : 1375065-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137153
Availability details for
Quantity
  • Availability
  • Add to Cart