ENA408616073 Enamine

5-(4-fluorophenyl)-4-methylthiophene-2-carboxamide

  • MDL number MFCD26407811
  • Cas Number : 1311587-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137166
Availability details for
Quantity
  • Availability
  • Add to Cart