ENA408616076 Enamine

4-chloro-3,8-dimethylquinoline

  • MDL number MFCD18448613
  • Cas Number : 39593-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133393

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133393
Availability details for
Quantity
  • Availability
  • Add to Cart