ENA408616087 Enamine

2-(4-hydroxyphenyl)-2-methoxyacetic acid

  • MDL number MFCD20658832
  • Cas Number : 90536-45-1
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-hydroxyphenyl)(methoxy)acetic acid
Inchi Code 1S/C9H10O4/c1-13-8(9(11)12)6-2-4-7(10)5-3-6/h2-5,8,10H,1H3,(H,11,12)
Inchi Key UHWCKIPLXDQZMD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137192
Availability details for
Quantity
  • Availability
  • Add to Cart