ENA408616089 Enamine

[2-(diethylamino)ethyl](propyl)amine

  • MDL number MFCD11144301
  • Cas Number : 90723-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137221
Availability details for
Quantity
  • Availability
  • Add to Cart