ENA408616099 Enamine

2-(2-fluoro-4-nitrophenyl)-2-methylpropanoic acid

  • MDL number MFCD19695856
  • Cas Number : 1314735-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133710
Availability details for
Quantity
  • Availability
  • Add to Cart