ENA408616106 Enamine

1-bromo-4-cyclopentylbutan-2-one

  • MDL number MFCD11656501
  • Cas Number : 1218765-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137240

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137240
Availability details for
Quantity
  • Availability
  • Add to Cart