ENA408616120 Enamine

6,6-dimethyl-3-(4-methylbenzoyl)-4,5,6,7-tetrahydro-1-benzothiophen-2-amine

  • MDL number MFCD26407605
  • Cas Number : 1461715-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133841
Availability details for
Quantity
  • Availability
  • Add to Cart