ENA408616122 Enamine

1,3-dibromo-4-iodo-5-methyl-2-nitrobenzene

  • MDL number MFCD11846028
  • Cas Number : 1160573-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137348
Availability details for
Quantity
  • Availability
  • Add to Cart