ENA408616126 Enamine

N1-[(4-chlorophenyl)methyl]benzene-1,2-diamine

  • MDL number MFCD11186882
  • Cas Number : 5729-18-0
  • Molecular Weight : 232.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137354
Availability details for
Quantity
  • Availability
  • Add to Cart