ENA408616135 Enamine

ethyl 4-tert-butyl-1,3-thiazole-2-carboxylate

  • MDL number MFCD14654431
  • Cas Number : 79247-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137373
Availability details for
Quantity
  • Availability
  • Add to Cart