ENA408616137 Enamine

4-(3-methoxyphenyl)pyrrolidin-2-one

  • MDL number MFCD09955049
  • Cas Number : 38175-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133889

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133889
Availability details for
Quantity
  • Availability
  • Add to Cart