ENA408616165 Enamine

N-(4-fluoro-3-methylphenyl)acetamide

  • MDL number MFCD20360301
  • Cas Number : 331-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137449
Availability details for
Quantity
  • Availability
  • Add to Cart