ENA408616168 Enamine

2,2-dimethyl-5-{[(pyridin-4-yl)amino]methylidene}-1,3-dioxane-4,6-dione

  • MDL number MFCD26401142
  • Cas Number : 643069-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134091

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134091
Availability details for
Quantity
  • Availability
  • Add to Cart