ENA408616172 Enamine

6-fluoro-2,3-dihydro-4H-thiochromen-4-one

  • MDL number MFCD00078488
  • Cas Number : 21243-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07762

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07762
Availability details for
Quantity
  • Availability
  • Add to Cart