ENA408616174 Enamine

5-bromo-2-methyl-1,3-benzoxazol-7-amine

  • MDL number MFCD16468566
  • Cas Number : 1226067-50-0
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-methyl-1,3-benzoxazol-7-amine
Inchi Code 1S/C8H7BrN2O/c1-4-11-7-3-5(9)2-6(10)8(7)12-4/h2-3H,10H2,1H3
Inchi Key VQWRZDSIMJRUCP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137461

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-137461
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart