ENA408616177 Enamine

3-hydroxy-3-methyl-1$l^{6}-thiolane-1,1-dione

  • MDL number MFCD18277940
  • Cas Number : 873-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137527

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137527
Availability details for
Quantity
  • Availability
  • Add to Cart