ENA408616180 Enamine

5-(2,5-dichlorophenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD05182402
  • Cas Number : 502685-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 269...271
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137537

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137537
Availability details for
Quantity
  • Availability
  • Add to Cart