ENA408616181 Enamine

N-propyl-3-(trifluoromethyl)-1H-pyrazol-5-amine

  • MDL number MFCD21602186
  • Cas Number : 1796915-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137544
Availability details for
Quantity
  • Availability
  • Add to Cart