ENA408616187 Enamine

6-(4-methylphenyl)-6-oxohexanoic acid

  • MDL number MFCD01320048
  • Cas Number : 100847-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134176

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134176
Availability details for
Quantity
  • Availability
  • Add to Cart