ENA408616188 Enamine

3,3-dimethyl-5-(4-methylphenyl)-5-oxopentanoic acid

  • MDL number MFCD17409527
  • Cas Number : 92864-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134208
Availability details for
Quantity
  • Availability
  • Add to Cart