ENA408616203 Enamine

2-chloro-6-[(4-chlorophenyl)sulfanyl]benzonitrile

  • MDL number MFCD01566283
  • Cas Number : 91822-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134352

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134352
Availability details for
Quantity
  • Availability
  • Add to Cart