ENA408616209 Enamine

ethyl 6-bromo-8-methylimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD19105297
  • Cas Number : 907945-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134425

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134425
Availability details for
Quantity
  • Availability
  • Add to Cart