ENA408616225 Enamine

4-(2-hydroxyethyl)benzene-1-sulfonamide

  • MDL number MFCD00837255
  • Cas Number : 829-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134525

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134525
Availability details for
Quantity
  • Availability
  • Add to Cart