ENA408616228 Enamine

1,3-dimethyl-5-(methylsulfanyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD18879342
  • Cas Number : 1340235-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134569

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134569
Availability details for
Quantity
  • Availability
  • Add to Cart