ENA408616235 Enamine

3-(morpholin-4-yl)pyridin-4-amine

  • MDL number MFCD20715635
  • Cas Number : 144864-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134596
Availability details for
Quantity
  • Availability
  • Add to Cart