ENA408616237 Enamine

tert-butyl N-[2-(pyridin-4-yl)-1,3-thiazol-4-yl]carbamate

  • MDL number MFCD24467297
  • Cas Number : 692889-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134601
Availability details for
Quantity
  • Availability
  • Add to Cart