ENA408616313 Enamine

2-(2-chloroacetamido)-3-methylbutanoic acid

  • MDL number MFCD00062945
  • Cas Number : 4090-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17602

Safety Information

signalword Warning
safety_statement 26-36
msds https://www.enaminestore.com/catalog/EN300-17602
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart