ENA408616380 Enamine

1-nitro-4-(4-nitrophenyl)benzene

  • MDL number MFCD00039744
  • Cas Number : 1528-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242...244
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18693

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18693
Availability details for
Quantity
  • Availability
  • Add to Cart