ENA408616577 Enamine

N-[(3,5-dimethoxyphenyl)methyl]aniline

  • MDL number MFCD01468696
  • Cas Number : 625407-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32386
Availability details for
Quantity
  • Availability
  • Add to Cart