ENA408616614 Enamine

3-(propan-2-yl)-6-(pyridin-4-yl)pyrido[3,2-d][1,2]oxazole-4-carboxylic acid

  • MDL number MFCD09737563
  • Cas Number : 954278-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56440
Availability details for
Quantity
  • Availability
  • Add to Cart