ENA408616620 Enamine

3-aminopentanenitrile

  • MDL number MFCD02101363
  • Cas Number : 75405-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64682
Availability details for
Quantity
  • Availability
  • Add to Cart