ENA408616625 Enamine

1-methyl-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD09065021
  • Cas Number : 1020721-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66957

Safety Information

signalword Warning
pcodes P261,P264,P272,P280,P302+P352,P305+P351+P338,P333+P313,P337+P313,P363,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-66957
pictograms GHS07
hstatements H317,H319
Availability details for
Quantity
  • Availability
  • Add to Cart