ENA408616636 Enamine

6-ethyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD03789268
  • Cas Number : 672904-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75773
Availability details for
Quantity
  • Availability
  • Add to Cart