ENA408616647 Enamine

2-(ethanesulfonyl)acetonitrile

  • MDL number MFCD11934304
  • Cas Number : 13654-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-95884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95884
Availability details for
Quantity
  • Availability
  • Add to Cart